Learn More
FyterTech Nonwovens Spilfyter™ HazMat Poly Drum Spill Kit
Spill response materials packaged in a Poly Drum
Supplier: FyterTech Nonwovens JV250055
Comprend:
3 in. x 4 ft. Socks (20), 16 x 18 in. Pads (200), Disposal Bag w/ Twist Tie (1), Nitrile Gloves (1 pr.), Splash Goggles (1 pr.), Emergency Response Guidebook (1), 55 Gallon Poly Drum (1)
Description
- Kits contained in durable Poly Drum with Lever-Lock metal ring tops
- Contains Hazmat spill response socks and pads for aggressive or unkown solution spills
- Designed for either inside or outside storage
- Absorbs up to 72 gal. (272 liters) per kit
Compliance
Poly Drums are UN-Certified
Spécifications
| 55 gal. | |
| Polypropylene Particulate, Fabric | |
| 20 socks, 200 pads, 1 twist tie, 1 pair nitrile gloves, 1 pair splash goggles, 1 emergency response guidebook, 1 poly drum 55 gal. | |
| 29 kg |
| 208 L | |
| Sorbent Spill Kit | |
| 64 lb. |