missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMB Chromogen Solution (for ELISA)

Catalog No. 002023
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
002023 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 002023 Supplier Invitrogen™ Supplier No. 002023

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

May be used as a substrate for horseradish peroxidase (HRP) in ELISAs

Use TMB (3,3', 5,5;-tetramethylbenzidine) chromogen solution as supplied. No dilution or further preparation is required. In the presence of peroxidase (HRP, POD), TMB will be converted to a blue color that can be read at OD650 nm.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

Specifications

Content And Storage Store at 2°C to 8°C.
Detection Method Colorimetric
Product Type TMB Solution
Substrate TMB
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Quantity 500 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.