missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Running Buffer Kit DFS Item

Contains the NativePAGE Running Buffer (20X)

Supplier:  Invitrogen™ BN2007

Numéro de catalogue. BN2007


Il en reste null

Comprend: 1L of NativePAGE Running Buffer (20X), 250mL NativePAGE Cathode Buffer Additive (20X)

Description

Description

NativePAGE Running Buffer (20X), is used to make the NativePAGE Cathode and Anode Running Buffers for use with an XCell SureLock™ Mini Cell when running NativePAGE Gels. The kit also contains the NativePAGECathode Buffer Additive (20X) that is used with the NativePAGE Running Buffer to make the Cathode Running Buffer for the system.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Spécifications

Spécifications

Contains:
• 1 L of NativePAGE™ Running Buffer (20X)
• 250 ml NativePAGE™ Cathode Buffer Additive (20X)
Store at room temperature.
Native-PAGE
1 kit
PAGE Running Buffer Kit
Running Buffers
NativePAGE™ Gels
NativePAGE
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.