missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lumio™ Green Detection Kit
Enables immediate visualization of Lumio™ fusion protein bands directly in a polyacrylamide gel
Supplier: Invitrogen™ LC6090
Comprend: The Lumio™ Green Detection Kit contains one vial of Lumio™ Green Detection Reagent, five vials of Lumio™ Gel Sample Buffer, and one vial of Lumio™ In-Gel Detection Enhancer.
Description
The Lumio™ Green Detection Kit is a sensitive and highly specific kit for labeling Lumio™ fusion proteins prior to electrophoresis.
- Label proteins fused to a Lumio sequence
- Visualize results immediately after electrophoresis, eliminate gel staining and western blotting
- Detect nanogram levels of protein with UV transilluminator, equipped with standard camera (Figure 1), or laser-based scanner
Enzyme and Protein Activity Assays, Lumio Detection, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Reporter Assays
Order Info
Shipping Condition: Dry Ice
Spécifications
| The Lumio™ Green Detection Kit contains one vial of Lumio™ Green Detection Reagent, five vials of Lumio™ Gel Sample Buffer, and one vial of Lumio™ In-Gel Detection Enhancer. Store at -20°C. All components are guaranteed stable for 6 months when properly stored. | |
| Fluorescence | |
| Pre-Gel Fusion Protein Labeling Kit | |
| 1 kit | |
| Proteins (Lumio™-Tagged) , Proteins (TC-Tagged) |
| In-Gel Detection | |
| Lumio Green | |
| Lumio | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.