missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Amyloid beta 42 ELISA Kit

Catalog No. KHB3441
Encompass
Change view
Click to view available options
Quantity:
2 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHB3441 96 Tests
KHB3442 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KHB3441 Supplier Invitrogen™ Supplier No. KHB3441
Only null left

Includes

Pre-coated plate, standard, detector antibody, anti-IgG-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

ELISA

The Human Amyloid Beta 42 (Hu Abeta 42) ELISA quantitates Hu Abeta 42 in human tissue culture medium, tissue homogenate, and cerebrospinal fluid (CSF). The assay will exclusively recognize both natural and recombinant Hu Abeta 42. Principle of the method The Human Abeta 42 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Beta amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein by a protease, gamma-secretase. Beta Amyloid (1-40) (Amyloid beta 40 or Abeta 40) together with beta Amyloid (1-42) (Amyloid beta 42 or Abeta 42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the Amyloid beta 40 and Amyloid beta 42 alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.

Order Info

Shipping condition: Wet ice

Specifications

Accession Number P05067
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity <10 pg/mL
Concentration 15.6 to 1000 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cerebrospinal Fluid, Supernatant, Tissue Homogenates
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 351
Gene Symbol AAA,ABETA,ABPP,AD1I,CTFgamma,CVAP,PN-II,PN2,APP
Incubation Time 4 hr.
Interassay CV 4.3%
Intraassay CV 3.6
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, Anti-IgG HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Aβ 42
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cerebrospinal Fluid 50 μL, Supernatant 50 μL, Tissue Homogenate 50 μL
Storage Requirements 2°C to 8°C
Target Amyloid beta 42
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.