Learn More
MilliporeSigma™ Novagen™ pSTBlue-1 AccepTor™ Vector Giga Cloning Kit
Features NovaBlue GigaSingles™ Competent Cells for increased transformation efficiency. MilliporeSigma™; Novagen™ pSTBlue-1 AccepTor™ Vector Giga Cloning Kit is designed for the cloning of PCR products generated using thermostable DNA polymerases that leave single 3'-dA nucleotide overhangs.
$659.00 - $1,080.00
Spécifications
| Product Type | pSTBlue-1 Vector Giga Cloning Kit |
|---|---|
| Content And Storage | Store at ≤70°C. |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
712283
|
MilliporeSigma™
712283 |
20 Reactions |
chaque for $659.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
712284
|
MilliporeSigma™
712284 |
40 Reactions |
chaque for $1,080.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Vector enables direct ligation by virtue of single 3'-dU overhangs that anneal with 3'-dA extensions on PCR products, the dU residues are converted to dT residues in vivo
- No restriction enzymes or special primers
- Direct ligation of PCR product with vector
- Compatible with polymerases that leave single 3´-dA overhangs
- Blue/white screening with pSTBlue-1 or pETBlue™-1 vectors
- Simple protocol takes as little as 40 minutes from PCR product to plating transformants
Spécifications
| pSTBlue-1 Vector Giga Cloning Kit | |
| Store at ≤70°C. |
For Research Use Only