Learn More
MilliporeSigma™ Novagen™ pSTBlue-1 AccepTor™ Vector Giga Cloning Kit
Description
- Vector enables direct ligation by virtue of single 3'-dU overhangs that anneal with 3'-dA extensions on PCR products, the dU residues are converted to dT residues in vivo
- No restriction enzymes or special primers
- Direct ligation of PCR product with vector
- Compatible with polymerases that leave single 3´-dA overhangs
- Blue/white screening with pSTBlue-1 or pETBlue™-1 vectors
- Simple protocol takes as little as 40 minutes from PCR product to plating transformants
Specifications
Specifications
| Product Type | pSTBlue-1 Vector Giga Cloning Kit |
| Content And Storage | Store at ≤70°C. |
| Quantity | 40 Reactions |
For Research Use Only