missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 262890050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC262890050


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Norethisterone
205.0°C to 206.0°C
Authentic
5 g
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dioxane
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
298.42
CHEBI:7627
≥97.5% (HPLC)
68-22-4
0.5% max. (1 g, 105°C, 3 h)
C20H26O2
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
VIKNJXKGJWUCNN-XGXHKTLJSA-N
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
6230
298.42

Chemical Identifiers

68-22-4
298.42
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
CHEBI:7627
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
C20H26O2
VIKNJXKGJWUCNN-XGXHKTLJSA-N
6230
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of causing cancer. May damage fertility. May damage the unborn child. May cause harm to breast-fed children.

GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention. Avoid contact during pregnancy/while nursing.

GHS Signal Word: Danger

EINECSNumber : 200-681-6

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only