missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Nitrogen Standard, 100 ppm N (328.5 ppm NO2-), Ricca Chemical

$233.30 - $429.67

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
54611-4
View Documents
Ricca Chemical Company
546114
120 mL Amber Glass Bottle
Each for $233.30
Only null left
 
54611-16
View Documents
Ricca Chemical Company
5461116
500 mL Amber Glass Bottle
Each for $429.30
Only null left
 
54611-32
View Documents
Ricca Chemical Company
5461132
1 L Amber Glass Bottle
Each for $429.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7758-09-0
85.10
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium nitrite
KNO2
MFCD00011408
516910
[K+].[O-]N=O

Specifications

Amber Glass Bottle
98.0,0.06
Liquid
MFCD00011408
Approximately 7
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium nitrite
120 mL
1mL = 0.1mg N, 100ppm N as Nitrite (328.5ppm NO2-)
7758-09-0,7732-18-5,7758-09-0
100.0,0.06
KNO2
Colorless
100°C
[K+].[O-]N=O
85.10
516910
Nitrogen Standard
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

SDS
missing translation for 'documents'

missing translation for 'documents'