missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrite Nitrogen Standard, 100 ppm N (328.5 ppm NO2-), Ricca Chemical
Supplier: Ricca Chemical Company 5461132
Specifications
| Nitrogen Standard | |
| 7758-09-0,7732-18-5,7758-09-0 | |
| 100.0,0.06 | |
| KNO2 | |
| Colorless | |
| 100°C | |
| [K+].[O-]N=O | |
| 85.10 | |
| 516910 |
| Amber Glass Bottle | |
| 98.0,0.06 | |
| Liquid | |
| MFCD00011408 | |
| Approximately 7 | |
| BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
| potassium nitrite | |
| 1 L | |
| 1mL = 0.1mg N, 100ppm N as Nitrite (328.5ppm NO2-) |
Chemical Identifiers
| 7758-09-0 | |
| 85.10 | |
| BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
| potassium nitrite |
| KNO2 | |
| MFCD00011408 | |
| 516910 | |
| [K+].[O-]N=O |
Safety and Handling
ShelfLife : 6 months