missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Nitrogen Standard, 100 ppm N (328.5 ppm NO2-), Ricca Chemical

Supplier:  Ricca Chemical Company 5461116

 View more versions of this product

Catalog No. 54611-16

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nitrogen Standard
7758-09-0,7732-18-5,7758-09-0
100.0,0.06
KNO2
Colorless
100°C
[K+].[O-]N=O
85.10
516910
Amber Glass Bottle
98.0,0.06
Liquid
MFCD00011408
Approximately 7
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium nitrite
500 mL
1mL = 0.1mg N, 100ppm N as Nitrite (328.5ppm NO2-)

Chemical Identifiers

7758-09-0
85.10
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium nitrite
KNO2
MFCD00011408
516910
[K+].[O-]N=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

SDS
Documents

Documents

Promotions

Promotions