missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocellulose Membranes, 0.2 μm

Numéro de catalogue. PI77012
Change view
Click to view available options
Dimensions (LxW):
10.5 x 7.9 cm
8 x 8 cm
12 x 8 cm
Sufficient For:
15 Mini-gel Blots
25 Mini-gel Blots
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Dimensions (LxW) Sufficient For
PI77012 12 x 8 cm 25 Mini-gel Blots
PI88013 10.5 x 7.9 cm 15 Mini-gel Blots
PI88024 8 x 8 cm 15 Mini-gel Blots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PI77012 Fournisseur Thermo Scientific™ Code fournisseur LSG77012
Il en reste null

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.2-μm pore size is well-suited for western blotting, and this smaller pore size is ideal for the transfer of low molecular weight proteins or peptides and nucleic acids (<300 bp).

Features include:
100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background—produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting

Spécifications

Length (Metric) 12 cm
Width (Metric) 8 cm
Description Pierce Nitrocellulose Membrane, 0.2 μm, 12 x 8 cm
Quantity 25 Pre-cut Membranes
Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage Store membranes flat at ambient temperature, away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. Keep membranes out of direct sunlight. Sunlight may cause the nitrocellulose to dry and become brittle. Note: Nitrocellulose membrane filters are highly flammable. Keep away from heat and open flame. Flash point is approximately 2°C. Membranes can be sterilized by steam-autoclaving at 121°C.
Shipping Condition Room Temperature
Dimensions (LxW) 12 x 8 cm
Sufficient For 25 Mini-gel Blots
Afficher plus de résultats Afficher moins de résultats