Learn More
Nicotinoyl chloride hydrochloride, 97%
CAS: 20260-53-1 | C6H4ClNO·HCl | 178.02 g/mol
$188.18 - $188.18
Identifiants chimiques
| CAS | 20260-53-1 |
|---|---|
| Molecular Formula | C6H4ClNO·HCl |
| Molecular Weight (g/mol) | 178.02 |
| MDL Number | MFCD00012820 |
| InChI Key | MSYBLBLAMDYKKZ-UHFFFAOYSA-N |
| Synonym | nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl |
| PubChem CID | 88438 |
| IUPAC Name | pyridine-3-carbonyl chloride;hydrochloride |
| SMILES | C1=CC(=CN=C1)C(=O)Cl.Cl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC338410250
|
Thermo Scientific Chemicals
338410250 |
25 g | Glass bottle |
chaque for $188.18
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 20260-53-1 | |
| 178.02 | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| 88438 | |
| C1=CC(=CN=C1)C(=O)Cl.Cl |
| C6H4ClNO·HCl | |
| MFCD00012820 | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| pyridine-3-carbonyl chloride;hydrochloride |
Spécifications
| 20260-53-1 | |
| 100.0 | |
| White | |
| 97% | |
| C6H4ClNO·HCl | |
| 25 g | |
| Solubility in water: decomposes | |
| C1=CC(=CN=C1)C(=O)Cl.Cl | |
| 178.02 | |
| 178.02 | |
| Crystalline Powder |
| 96.0 | |
| 151.0°C to 156.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00012820 | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| pyridine-3-carbonyl chloride;hydrochloride | |
| 88438 | |
| 97% | |
| Nicotinoyl chloride hydrochloride, 97% |
Sécurité et manipulation
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 243-648-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only