missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat NGF beta-Cell Lysates ELISA Kit

Numéro de catalogue. ERNGFCL
Encompass
Modifier l'affichage
Click to view available options
Quantité:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
ERNGFCL 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. ERNGFCL Fournisseur Invitrogen™ Code fournisseur ERNGFCL

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

ELISA

The Rat Nerve Growth Factor Beta-Cell Lysates (Rt NGFbeta) ELISA quantitates Rt NGFbeta in rat cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Rt NGFbeta. Principle of the method The Rat NGFbeta solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.

Spécifications

Numéro d'enregistrement P25427
Plage de l'essai 15-15000 pg/mL
Sensibilité d’essai 15 pg/mL
Plage d’étalonnage 15 pg/mL to 15000 pg/mL
Contenu et stockage Pre-coated 96-well plate, standard, detector antibody, streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. May be stored at 2–8°C for up to 6 months from the date of shipment.
Essai Solid-phase sandwich ELISA
Conjugué HRP
Format de la trousse d’immunodosage Sandwich ELISA Kit
Type de produit ELISA
Famille de protéines Growth Factors & Receptors
Type d’échantillon Cell Lysates, Plasma, Serum, Supernatant, Tissue Homogenates
Spécificité This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, TIMP-1, TNF-alpha.
Suffisant pour 96 assays
Espèces cibles Rat
À utiliser avec (application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
À utiliser avec (équipement) Absorbance Microplate Reader
Alias du gène B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGFB nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF; Ngfb; beta-NGF
ID du gène (Entrez) 310738
Symbole de gène NGF
CV intra-essai <12%, <10%
Contenus des kits Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell Lysis Buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell Lysis Buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Étiquette ou colorant Biotin
Alias des protéines Beta-NGF
Quantité 96 Tests
Statut réglementaire RUO
Disciplines de recherche Immunology
Volume d’échantillon Cell Lysate 20 μL
Shipping condition (Condition d'expédition) Wet or Dry Ice
Conditions de stockage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonyme Tgfb
Cible NGF
Durée du test 1 hr 20 min
Durée totale de l'essai 4 hr 45 min
Afficher plus de résultats Afficher moins de résultats