missing translation for 'onlineSavingsMsg'
Learn More

Sartorius NexaGel ORGANOID

Numéro de catalogue. 14558133
Modifier l'affichage
Click to view available options
Product Type:
Discovery Kit
Organoid 1
Organoid 3
Organoid 4
Organoid-2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type
14558133 Organoid 4
14558130 Discovery Kit
14558129 Organoid 1
14558131 Organoid-2
14558132 Organoid 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 14558133 Fournisseur Sartorius Code fournisseur NGH044

Animal origin-free hydrogels specifically designed to support the cultivation of patient-derived organoids.

  • NexaGel™ ORGANOID (1-4) are animal origin-free hydrogels specifically designed to support the cultivation of patient-derived organoids, organoids from pluripotent stem cells (PSCs), co-cultures, and PDX models.
  • The NexaGel™ ORGANOID KIT includes four unique hydrogels, each with varying bio-functional ligands, mechanical properties, and biodegradability to cater to diverse organoid culture needs.
  • These hydrogels are ready for immediate use at room temperature, maintaining a neutral pH, transparency, permeability, and compatibility with various imaging systems.
  • They form a hydrogel matrix simply by mixing with the cell culture medium.
  • NexaGel™ ORGANOID hydrogels are versatile, suitable for both 3D cell culture and 2D hydrogel coating applications.
  • Moreover, these hydrogels can be used alongside NexaGel™ STEM (Cat# NGH02), a hydrogel system designed for 3D static suspension cultures and the expansion of human pluripotent stem cells.
  • Stem cell spheroids cultured in NexaGel™ STEM can be transferred to NexaGel™ ORGANOID hydrogels for further organoid differentiation.
  • Essential growth factors and molecules can be incorporated directly into the hydrogel matrix or applied on top.
  • Organoids grown in this system can be easily harvested using the NexaGel™ Cell Recovery Solution.
  • NexaGel™ ORGANOID hydrogels offer a well-defined 3D microenvironment, advancing the field of personalized medicine.

Spécifications

Cell Type Organoid
Form Liquid
Product Type Organoid 4
Quantity 10 mL