missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neurobasal™-A Medium, minus phenol red
GREENER_CHOICE

Catalog No. 12349015
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12349015 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12349015 Supplier Gibco™ Supplier No. 12349015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Neurobasal™-A Medium, minus phenol red

Neurobasal™-A Medium Minus Phenol Red is a basal media that meets the special requirements of post-natal and adult brain neuronal when used with GIBCO™ B-27™ Supplement, but without Phenol Red. Neurobasal™-A Medium can be used to grow neuronal cells from hippocampus, cortex and other regions of the brain. Neurobasal™-A Medium allows for both long and short term maintenance of homogeneous populations of neuronal cells without the need of an astrocyte feeder layer.

Use Neurobasal™-A Medium Minus Phenol Red for applications in which the presence of Phenol Red will interfere with your results.

Add Your Own Supplements
All Neurobasal™ media contain no L-glutamine, L-glutamic acid, or aspartic acid. Neurobasal™ media must be supplemented with 0.5 mM L-glutamine or GlutaMAX™-I Supplement and either a serum-free supplement (such as B-27™ Supplement or N-2 Supplement), or with serum. For initial plating, you should add 25μM glutamic acid.

Quality Control Testing
All Neurobasal™ media are tested for pH, osmolality, endotoxin, and the absence of bacterial and fungal contamination.

We also provide Neurobasal™ -A Medium with Phenol Red.

Neurobasal™-A Medium is optimized for post-natal and adult brain neurons. For growing pre-natal and embryonic neurons, we provide Neurobasal™ Medium.

See the entire Neurobasal™ product line.

Learn more about neural cell culture products from Invitrogen at www.invitrogen.com/neuralculture.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type Neurobasal Medium
Without Additives No Phenol Red
Quantity 500 mL
Shipping Condition Ambient
Tested For Endotoxin, Performance, Sterility, Osmolality, pH
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Neuronal Cell Culture
Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?

No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.

What supplements should I use with Neurobasal or Neurobasal-A medium?

Neurobasal and Neurobasal-A media (for postnatal and adult neurons) allow for long-term maintenance of neuronal cells without the need for an astrocyte feeder layer. These media should be supplemented with either serum or a serum-free supplement, plus 0.5mM L-glutamine. B-27 supplement is a serum-free supplement that comes as a 50X concentrate in a 10ml volume. This is enough supplement for 500ml of media. Fetal, postnatal, and adult neural cultures can be grown in the appropriate Neurobasal medium supplemented with B-27 supplement .

We also have two other supplements. One is called G-5 and is for growth and expression of glial cells (normal and tumor) of astrocytic phenotype. This comes in a 1ml size, at a 100X concentration. The other supplement is called N-2 and is for growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype. This comes in a 5ml size, at a 100X concentration.

For more information on Neurobasal media, search "Neurobasal" from our website home page.

Can you provide references for growing adult neurons?

Instructions for growing adult neurons can be found in the Neurobasal-A Medium product insert. You can also refer to the following publication for growing adult neurons:
• Isolation and Culture of Adult Rat Hippocampal Neurons by Brewer, G.J., J. Neuroscience Methods 71 (1997), 145-158.

Should I use Neurobasal-A Medium with B27 and b-FGF for postnatal neurons?

We do not know at what age postnatal neurons require FGF and a higher osmolality medium, or how this could vary based on the species from which the neural tissue is isolated. Without this knowledge, we generally recommend switching to the Neurobasal-A Medium for isolates from animals above 1 week of age. You will need to determine if the Neurobasal-A Medium is optimal for neurons you have from one day old rats.

Further, the addition of bFGF can increase the viability of neurons of any age. Therefore, caution needs to be made in impure neuronal cell culture as bFGF can increase the proliferation of non-neuronal cells. We recommend a pilot experiment to test the impure culture in the presence and absence of bFGF.

What should I add to Neurobasal-A Medium to grow adult neurons?

We recommend adding B27 and b-FGF to Neurobasal-A Medium to grow adult neurons.

What did you add to Neurobasal-A Medium to increase osmolality?

4 g/L of NaCl has been added to the Neurobasal-A Medium formulation to increase osmolality.

I understand that some media are worse than others for fluorescence imaging. How do I choose?

Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.

Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?

Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.

Why is the color of Neurobasal Medium without Phenol red yellow?

The yellow color comes from vitamin components.

Which supplement can I add to Neurobasal Medium to culture my neural cells?

As a general guideline, we recommend using Gibco B-27 Plus Supplement to culture neural stem cells, hippocampal, and other CNS neurons.

- For neuroblastomas, or post-mitotic neurons from both PNS and CNS, Gibco N-2 Supplement can be used.
- For primary glial cells (astrocytes) or tumor cell lines of astrocytic phenotype (astrocytes and gliomas), or oligodendrocytes, Gibco G-5 Supplement can be used.


For Research Use Only. Not for use in diagnostic procedures.