Learn More
Neodymium(III) nitrate hexahydrate, 99.9% (REO)
CAS: 16454-60-7 | H12N3NdO15 | 438.344 g/mol
$148.12 - $148.12
Identifiants chimiques
| CAS | 16454-60-7 |
|---|---|
| Molecular Formula | H12N3NdO15 |
| Molecular Weight (g/mol) | 438.344 |
| MDL Number | MFCD00149799 |
| InChI Key | VQVDTKCSDUNYBO-UHFFFAOYSA-N |
| Synonym | neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate |
| PubChem CID | 204494 |
| IUPAC Name | neodymium(3+);trinitrate;hexahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1291218
|
Thermo Scientific Chemicals
01291218 |
50 g |
chaque for $148.12
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Neodymium(III) nitrate hexahydrate is used to prepare nanoparticles of neodymium oxybromide and neodymium oxide. It is also used in coloring glasses and for doping some glass lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16454-60-7 | |
| 438.344 | |
| VQVDTKCSDUNYBO-UHFFFAOYSA-N | |
| 204494 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
| H12N3NdO15 | |
| MFCD00149799 | |
| neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate | |
| neodymium(3+);trinitrate;hexahydrate |
Specifications
| ∼40°C | |
| 3.0 to 4.0 | |
| 50 g | |
| MFCD00149799 | |
| 14,6450 | |
| Soluble in water and alcohol. | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] | |
| 438.344 | |
| 438.34 (330.25 Anhydrous) | |
| Odorless | |
| Neodymium(III) nitrate hexahydrate |
| 16454-60-7 | |
| Crystalline | |
| H12N3NdO15 | |
| UN1477 | |
| neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate | |
| VQVDTKCSDUNYBO-UHFFFAOYSA-N | |
| neodymium(3+);trinitrate;hexahydrate | |
| 204494 | |
| 99.9% | |
| Hygroscopic | |
| (REO) |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P280-P370+P378q-P501c
H272
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 233-153-9
RTECSNumber : OJ6125000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only