Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure
GREENER_CHOICE
Store pastes, solids, specimens and lab supplies in these translucent, wide-mouth, straight-sided jars that offer excellent chemical resistance.
$123.93 - $325.29
Specifications
Color Translucent
Includes Polypropylene Caps
Material PPCO
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
1181510G
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21189050
15 mL 38 mm
Pack of 12 Each for $123.93
Case of 6 Pack for $743.58
Only null left
1181510A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180001
30 mL 43 mm
Pack of 12 Each for $134.75
Case of 6 Pack for $808.50
Only null left
1181510B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180002
60 mL 53 mm
Pack of 12 Each for $134.43
Case of 4 Pack for $537.72
Only null left
1181510C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180004
125 mL 70 mm
Pack of 12 Each for $225.14
Case of 3 Pack for $675.42
Only null left
1181510D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180008
250 mL 70 mm
Pack of 6 Each for $152.61
Case of 6 Pack for $915.66
Only null left
1181510E
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180016
500 mL 120 mm
Pack of 6 Each for $228.89
Case of 4 Pack for $915.56
Only null left
1181510F
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21180032
1000 mL 120 mm
Pack of 6 Each for $325.29
Case of 4 Pack for $1,301.16
Only null left
Description
Description
Store pastes, solids, specimens and lab supplies in Thermo Scientific™ Nalgene™ Wide-Mouth Straight-Sided PPCO Jars. These versatile, translucent jars have excellent chemical resistance and can be autoclaved repeatedly. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. Not liquid leakproof.
Features:
  • Wide mouth and straight sides provide full-access and complete sample retrieval.
  • 15, 30 and 60mL jars have natural polypropylene closures.
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking, which helps with organizing the fridge or counter top and maximizing storage space
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads; recommended for use with Fluid Transfer Closure DS2153-0700.
  • For 500mL and 1L sizes, remove closure and autoclave separately.
  • Polypropylene copolymer jars are not liquid leakproof, not recommended for liquid transport.
Recycling Information
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Specifications
Translucent
Polypropylene Caps
PPCO