Learn More
Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
Supplier: Thermo Scientific™ 21100006
Description
Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Features:- Resistant to acids, alcohols and bases
- Contact-clear bottle provides visible liquid level
- Square design provides a large area for labeling
- Linerless polypropylene closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
- Autoclavable
Note:
Completely disengage threads or remove cap before autoclaving.
Identifiants chimiques
| Translucent | |
| PPCO | |
| Yes | |
| Wide | |
| Square Wide Mouth Bottle | |
| No |
| Wide-Mouth Square Bottle | |
| Non-sterile | |
| No | |
| Square | |
| Unlined | |
| Autoclavable |
Spécifications
| 6 oz., 175 mL | |
| 6 oz. | |
| Wide-Mouth Square Bottle | |
| 1.1 in. | |
| PPCO | |
| 12 | |
| Natural, translucent | |
| Yes | |
| Wide | |
| 102 mm | |
| 2.05 in. | |
| Unlined | |
| 38 mm | |
| Screw Cap | |
| Autoclavable |
| 175 mL | |
| Translucent | |
| 2 in. | |
| 28 mm | |
| 72 | |
| Non-sterile | |
| USP Class VI; FDA - 21 CFR | |
| No | |
| Square | |
| Square Wide Mouth Bottle | |
| 4 in. | |
| Polypropylene | |
| 51 mm | |
| No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.