Learn More
Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
GREENER_CHOICE
Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
$337.79 - $1,351.55
Specifications
Color Translucent
Description Wide-Mouth Square Bottle
Material PPCO
Sterility Non-sterile
Closure Included Yes
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
033112A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21100002
60 mL 28 mm
Case of 72 Each for $593.34
Only null left
033112B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21100006
175 mL 38 mm
Case of 72 Each for $951.21
Only null left
033112C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21100008
250 mL 43 mm
Case of 72 Each for $1,351.55
Only null left
033112D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21100016
500 mL 53 mm
Pack of 12 Each for $337.79
Case of 4 Pack for $1,351.16
Only null left
033112E
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21100032
1 L 63 mm
Case of 24 Each for $982.45
Only null left
Description
Description
Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable
  • Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials
Note:
Completely disengage threads or remove cap before autoclaving.
.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifications
Specifications
Translucent
PPCO
Yes
Wide
Square Wide Mouth Bottle
No
Wide-Mouth Square Bottle
Non-sterile
No
Square
Unlined
Autoclavable
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.