Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
Supplier: Thermo Scientific™ 20190060
Description
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
Features:
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Suitable for frozen storage down to -40°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Sterile and non-pyrogenic
- Available in low particulate format
- Supported by extensive validation documentation
Ordering Information:
- Packed in shrink-wrapped trays
Note:
- 2 L size has molded-in handgrips
Warranty:
- 90 days
Identifiants chimiques
| Clear | |
| PETG | |
| -70°C to 70°C | |
| Yes | |
| Narrow | |
| Media Bottle | |
| Not Autoclavable |
| Media/Buffer Preparation , Media/Buffer Storage, Storage | |
| Sterile | |
| Bottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines. | |
| Yes | |
| Square | |
| HDPE | |
| Upstream media/buffer prep and storage. Upstream process solution storage. Upstream Validation |
Spécifications
| 60 mL | |
| Clear | |
| 0.71 in. | |
| Media/Buffer Preparation , Media/Buffer Storage, Storage | |
| 96/Cs. | |
| Sterile | |
| Bottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines. | |
| Yes | |
| Square | |
| Media Bottle | |
| HDPE | |
| 41 mm | |
| Upstream media/buffer prep and storage. Upstream process solution storage. Upstream Validation |
| 2 oz. | |
| 1.6 in. | |
| 18 mm | |
| PETG | |
| 24/Pk. | |
| -70°C to 70°C | |
| Yes | |
| Narrow | |
| 82 mm | |
| 3.22 in. | |
| 24 mm | |
| Not Autoclavable |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.