Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys

Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys are sterile, ready-to-use and designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
Supplier: Thermo Scientific™ 303042
Description
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
- Durable, translucent PC construction ensures the integrity of your sample
- Closures feature a silicone liner for leakproof† assurance
- Customization options are available for fluid transfer applications
- Extensive validation documentation support
- Suitable for use in storage conditions of -100° to +100°C
- Not for use on hot plates
Warranty: 90 days
Identifiants chimiques
| Polycarbonate | |
| -100°C to 100°C | |
| USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic | |
| Yes | |
| Storage Bottle | |
| Not Autoclavable |
| Sterile | |
| Silicone | |
| Yes | |
| Narrow | |
| Polypropylene | |
| Upstream process solution storage. Upstream Validation |
Spécifications
| 125 mL | |
| Clear | |
| 26.7 mm | |
| Polycarbonate | |
| 5/Pk. | |
| -100°C to 100°C | |
| USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic | |
| Yes | |
| Square | |
| Storage Bottle | |
| Polypropylene | |
| 52.1 mm | |
| Upstream process solution storage. Upstream Validation |
| 4.2 oz. | |
| 1.06 in. | |
| Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation | |
| 50/Cs. | |
| Sterile | |
| Silicone | |
| Yes | |
| Narrow | |
| 104.6 mm | |
| 4.1 in. | |
| 38 mm | |
| Not Autoclavable |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.