Learn More
Thermo Scientific™ Nalgene™ Dropper Bottles

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.
Supplier: Thermo Scientific™ 27519025
Description
Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.
- Dropper control tip snaps into place for a secure fit
- Dispensing tip and closures included, choose from white or multi-colored caps
- Leakproof†
Identifiants chimiques
| White | |
| Dispensing tip and closures | |
| 25 | |
| Yes | |
| Round | |
| 15-415 mm | |
| Yes |
| LDPE Dropper Bottle | |
| LDPE | |
| 25 | |
| 40 μL | |
| Dropping Bottle | |
| Dropper Control Tip |
Spécifications
| 8 mL | |
| White | |
| Dispensing tip and closures | |
| 25 | |
| White | |
| 8 mL | |
| Round | |
| 15-415 mm | |
| Yes |
| 0.27 oz. | |
| LDPE Dropper Bottle | |
| LDPE | |
| 25 | |
| Yes | |
| 40 μL | |
| Dropping Bottle | |
| Dropper Control Tip |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.