missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Dropper Bottles
GREENER_CHOICE

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.

$182.16 - $223.75

Specifications

Color White
Description LDPE Dropper Bottle
Includes Dispensing tip and closures
Material LDPE
No. per Case 25
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Closure Color Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Color Price Quantity & Availability  
0300717A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
27519125
4 mL White
Case of 25 Each for $182.16
Only null left
 
0300717B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
27519025
8 mL White
Case of 25 Each for $201.65
Only null left
 
0300719C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
27539050
15 mL Assorted
Case of 25 Each for $223.75
Only null left
 
0300717C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
27519050
15 mL White
Case of 25 Each for $204.51
Only null left
 
Description

Description

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Opaque white bottle ideal for UV lightsensistive solutions
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; choose white or multi-colored closures
  • Bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time
  • Leakproof† when used with provided Nalgene tip and closure

Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Specifications

Specifications

White
Dispensing tip and closures
25
Yes
Round
15-415 mm
Yes
LDPE Dropper Bottle
LDPE
25
40 μL
Dropping Bottle
Dropper Control Tip
Videos
SDS
Documents

Documents

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.