missing translation for 'onlineSavingsMsg'
Learn More

N-Bromophthalimide, 98+%

CAS: 2439-85-2 | C8H4BrNO2 | 226.029 g/mol

Supplier:  Thermo Scientific Chemicals A1160114

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1160114


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Brominating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

N-Bromophthalimide
204°C to 208°C
MFCD00005888
131211
n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
C1=CC=C2C(=C1)C(=O)N(C2=O)Br
226.029
226.03
2439-85-2
C8H4BrNO2
25 g
Light and moisture sensitive
MARXMDRWROUXMD-UHFFFAOYSA-N
2-bromoisoindole-1,3-dione
75542
≥98%

Identifiants chimiques

2439-85-2
226.029
MARXMDRWROUXMD-UHFFFAOYSA-N
2-bromoisoindole-1,3-dione
C8H4BrNO2
MFCD00005888
75542
C1=CC=C2C(=C1)C(=O)N(C2=O)Br
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 219-467-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only