missing translation for 'onlineSavingsMsg'
Learn More

N-(3-hydroxypropyl)phthalimide, 99%

CAS: 883-44-3 | C11H11NO3 | 205.21 g/mol

$94.75 - $94.75

Identifiants chimiques

CAS 883-44-3
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00023097
InChI Key BSMILTTURCQDGJ-UHFFFAOYSA-N
Synonym n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
PubChem CID 70160
IUPAC Name 2-(3-hydroxypropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC377010250
FDS Afficher les documents
Thermo Scientific Chemicals
377010250
25 g Glass bottle
chaque for $94.75
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

883-44-3
205.21
BSMILTTURCQDGJ-UHFFFAOYSA-N
70160
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO
C11H11NO3
MFCD00023097
n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
2-(3-hydroxypropyl)isoindole-1,3-dione

Spécifications

883-44-3
100.0
White
98.5% min. (GC)
C11H11NO3
25 g
Solubility in water: < 1g/l
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO
205.21
205.21
Crystalline Powder
98.5
75°C to 77°C
Authentic
Glass bottle
MFCD00023097
n-3-hydroxypropyl phthalimide, 2-3-hydroxypropyl isoindoline-1,3-dione, n-3-hydroxypropyl-phthalimide, 1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl, 3-phthalimido-1-propanol, 2-3-hydroxypropyl isoindole-1,3-dione, 2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione, 3-phthalimidopropanol, 3-phthalimidylpropanol, acmc-209qsg
BSMILTTURCQDGJ-UHFFFAOYSA-N
2-(3-hydroxypropyl)isoindole-1,3-dione
70160
99%
N-(3-Hydroxypropyl)phthalimide
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 212-931-1

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only