Learn More
Aqua Phoenix™ Five-in-One Test Strips
These multi-parameter test strips test for five water quality measurements on 1 strip.
Supplier: Aqua Phoenix™ EPD2755250
Description
These multi-parameter test strips test for five water quality measurements on 1 strip.
Great for quick, on-site evaluations of water quality with reliable results on every test. These mulit-parameter test strips test for five water quality measurements on 1 strip. Contains 50 tests.- Free Chlorine (0-10 mg/L)
- Total Chlorine (0-10 mg/L)
- Total Hardness (as CaCO3, 0-25 gpg and 0-425 mg/L)
- Total Alkalinity (as CaCO3, 0-240 mg/L)
- pH (6.2 - 8.4)
Specifications
| Free Chlorine (0-10 mg/L), Total Chlorine (0-10 mg/L), Total Hardness (as CaCO3, 0-25 gpg and 0-425 mg/L), Total Alkalinity (as CaCO3, 0-240 mg/L), pH (6.2 - 8.4) | |
| Test Strip | |
| 50 |
| 6.2 to 8.4 | |
| Water Quality Testing | |
| 5-in-1 Test Strip |