Learn More
MP Biomedicals™ LargeFlex™ Adapter for 2 x 250 mL Tube Holder on FastPrep 96
LargeFlex™ Adapter on FastPrep 96 holds 2 x 250 mL Lysing Matrix Bottles. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix bottles that remain securely sealed during the homogenization. For scale-up and small production run capabilities.
Supplier: MP Biomedicals™ 116010590
Description
- Product Overview: Super-sized adapter for the FastPrep-96™ that holds 2 x 250 mL Lysing Matrix Bottles. For scale-up and small production run capabilities. Compatible with the SafTest™ Food Inspection Platform. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix bottles that remain securely sealed during the homogenization.
- Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- A wide range of disposable 250 mL Lysing Matrix bottles ensure thorough homogenization of any sample type in seconds.
- Capacity: 2 tubes
Spécifications
| FastPrep-96 Adapters | |
| Nucleic Acid Isolation and Purification | |
| FastPrep-96 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.