missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse GM-CSF Quantikine ELISA Kit

Catalog No. MGM00
Encompass
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MGM00 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MGM00 Supplier R&D Systems™ Supplier No. MGM00

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse GM-CSF in cell culture supernates (50 ul), serum (50 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 5.8pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and recombinant mouse GM-CSF
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 12981
Quantity 2 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Synonym colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Show More Show Less