missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
$89.07 - $1,366.15
Specifications
| Color | Clear |
|---|---|
| For Use With (Application) | Ensures positive sample identification |
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Packaging | Shape | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Packaging | Shape | Price | Quantity & Availability | ||
|
14222211
|
Axygen™
MTS8CPC |
8 Cap Strip | Non-sterile | 120 Strips/PK., 5 Packs/Cs. |
Case of 600 Each for $275.48
|
|
|||
|
14222204
|
Axygen™
MTS118CRS |
1.1 mL | Sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Case of 600 Each for $1,232.22
|
|
|||
|
14222209
|
Axygen™
MTS12CPC |
12 Cap Strip | Non-sterile | 80 Strips/PK., 5 Packs/Cs. |
Case of 400 Each for $270.16
|
|
|||
|
14222200
|
Axygen™
MTS1112CR |
1.1 mL | Non-sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Case of 400 Each for $1,038.99
|
|
|||
|
14222201
|
Axygen™
MTS1112CRS |
1.1 mL | Sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Case of 400 Each for $1,366.15
|
|
|||
|
14222207
|
Axygen™
MTS11CRS |
1.1 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Pack of 960 Each for $243.78
Case of 5 Pack for $1,218.90
|
|
|||
|
14222212
|
Axygen™
MTS8CPCS |
8 Cap Strip | Sterile | 120 Strips/PK., 5 Packs/Cs. |
Case of 600 Each for $381.61
|
|
|||
|
14222197
|
Axygen™
MTS06CR |
0.65 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Case of 960 Each for $1,009.00
|
|
|||
|
14222210
|
Axygen™
MTS12CPCS |
12 Cap Strip | Sterile | 80 Strips/PK., 5 Packs/Cs. |
Case of 400 Each for $384.62
|
|
|||
|
14222198
|
Axygen™
MTS06CRS |
0.65 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Pack of 960 Each for $256.84
Case of 5 Pack for $1,284.20
|
|
|||
|
14222199
|
Axygen™
MTS1112C |
1.1 mL | Non-sterile | 80 Strips/Bag; 5 Bags/Cs. |
Case of 400 Each for $438.30
|
|
|||
|
14222203
|
Axygen™
MTS118CR |
1.1 mL | Non-sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Pack of 120 Each for $199.27
Case of 5 Pack for $996.35
|
|
|||
|
14222206
|
Axygen™
MTS11CR |
1.1 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. |
Pack of 960 Each for $207.81
Case of 5 Pack for $1,039.05
|
|
|||
|
14222202
|
Axygen™
MTS118C |
1.1 mL | Non-sterile | 120 Strips/Bag; 5 Bags/Cs. |
Case of 600 Each for $441.39
|
|
|||
|
14222196
|
Axygen™
MTS06C |
0.65 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. |
Case of 4800 Each for $437.26
|
|
|||
|
14222205
|
Axygen™
MTS11C |
1.1 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. |
Pack of 960 Each for $89.07
Case of 5 Pack for $445.35
|
|
|||
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
- Rack features a one-way lid design and alpha-numeric grid
- Molded-in condensation rings on the lid minimize cross contamination
Specifications
| Clear | |
| Ensures positive sample identification |