Learn More
Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
Supplier:  Axygen™ MTS1112C
 View more versions of this product
Catalog No. 14222199
Only null left
Description
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
Specifications
Chemical Identifiers
Clear
Ensures positive sample identification
Specifications
Autoclavable
Clear
Bag
Alpha numeric grid, one way lid
80 Strips/Bag; 5 Bags/Cs.
Yes
Microtube
1.1 mL
Polypropylene
Ensures positive sample identification
80/Pk.
Non-sterile
Round
Product Suggestions
Product Suggestions