missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard CE™ Hoods
MicroGuard CE™ Hoods, disposable protective clothing designed with your comfort in mind. Made from high MVTR, innovative fabric.
$314.35 - $314.35
Spécifications
| Product Type | Open Face |
|---|---|
| Size | Universal |
| Material | Microporous |
| Color | White |
| Closure Type | Elastic |
| Numéro de catalogue | Numéro du manufacturier. | Packaging Type | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Packaging Type | Prix | Quantité | |||||
|
19-472-564
|
International Enviroguard™
CE8063CI |
Individually Packaged |
caisse de 100 chaque for $314.35
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Microporous, clean processed
- Fabric keeps dust and liquids out while allowing air to flow freely
- Ideal for non-hazardous work environments
- Provide reliable liquid and particulate protection
- Tunnelized elastic offer secure closure and stronger seams
- Bulk or individually packaged to help prevent cross-contamination
- Environmentally friendly, can be recycled after use
Spécifications
| Open Face | |
| Microporous | |
| Elastic | |
| 100/Cs. | |
| Clean Processed |
| Universal | |
| White | |
| Serged | |
| Low linting, liquid barrier, tunnelized elastic face |