missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate, 98%, Spectrumâ„¢ Chemical

$336.44 - $20047.85

Chemical Identifiers

CAS 93-58-3
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611692
View Documents
Spectrum Chemical Mfg Cor
M1243500MLGL
500 mL Amber Glass Bottle
Each for $336.44
Only null left
 
18611691
View Documents
Spectrum Chemical Mfg Cor
M12434LTGL
4 L Amber Glass Bottle
Each for $1,185.21
Only null left
 
18611689
View Documents
Spectrum Chemical Mfg Cor
M124320LT
20 L Steel Can
Each for $3,042.06
Only null left
 
18611690
View Documents
Spectrum Chemical Mfg Cor
M1243200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

93-58-3

Specifications

1
1
1.514-1.518
1.082 - 1.088
Reagent
93-58-3
Amber Glass Bottle
500 mL
0.98
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.