accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 93-58-3

IUPAC Name: methyl benzoate
Synonyms: methyl benzoate
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: QPJVMBTYPHYUOC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (9)
Percent Purity 
  • (2)
  • (4)
  • (6)
Physical Form 
  • (12)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (4)
  • (2)
  • (6)
  • (3)
  • (9)
  • (2)
  • (4)
  • (6)
  • (12)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)