missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Benzoate, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M12434LTGL
Specifications
| 1 | |
| 1 | |
| 1.514-1.518 | |
| 1.082 - 1.088 | |
| Reagent |
| 93-58-3 | |
| Amber Glass Bottle | |
| 4 L | |
| 0.98 |
Chemical Identifiers
| 93-58-3 |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.