missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Benzoate, 98%, Spectrum™ Chemical
Specifications
Specifications
| Acidity | 1 |
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.514-1.518 |
| Quantity | 500 mL |
| Specific Gravity | 1.082 - 1.088 |
| Molecular Weight (g/mol) | 136.14 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Physical Form | Liquid |
| Show More |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.