missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M124320LT

 View more versions of this product

Catalog No. 18611689

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1
1
1.514-1.518
1.082 - 1.088
Reagent
93-58-3
Steel Can
20 L
0.98

Chemical Identifiers

93-58-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.