missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate, 98%, Spectrum™ Chemical

Catalog No. 18611690
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611690 200 L Drum
18611692 500 mL Amber Glass Bottle
18611691 4 L Amber Glass Bottle
18611689 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611690 Supplier Spectrum Chemical Mfg Cor Supplier No. M1243200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93-58-3

Specifications

Acidity 1
CAS Max % 1
Packaging Drum
Refractive Index 1.514-1.518
Quantity 200 L
Specific Gravity 1.082 - 1.088
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.