Learn More
Methyl 4-chloroacetoacetate, 97+%
CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol
$92.32 - $241.85
Identifiants chimiques
| CAS | 32807-28-6 |
|---|---|
| Molecular Formula | C5H7ClO3 |
| Molecular Weight (g/mol) | 150.56 |
| MDL Number | MFCD00000938 |
| InChI Key | HFLMYYLFSNEOOT-UHFFFAOYSA-N |
| Synonym | methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 |
| PubChem CID | 36240 |
| IUPAC Name | methyl 4-chloro-3-oxobutanoate |
| SMILES | COC(=O)CC(=O)CCl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAH6470914
|
Thermo Scientific Chemicals
H6470914 |
25 g |
chaque for $92.32
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6470922
|
Thermo Scientific Chemicals
H6470922 |
100 g |
chaque for $241.85
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 32807-28-6 | |
| 150.56 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| 36240 | |
| COC(=O)CC(=O)CCl |
| C5H7ClO3 | |
| MFCD00000938 | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| methyl 4-chloro-3-oxobutanoate |
Spécifications
| 32807-28-6 | |
| 1.305 g/mL | |
| 102°C (216°F) | |
| 1.456 | |
| 25 g | |
| 1759939 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| methyl 4-chloro-3-oxobutanoate | |
| 36240 | |
| ≥97% | |
| Methyl 4-chloroacetoacetate |
| Colorless | |
| 85°C (4 mmHg) | |
| C5H7ClO3 | |
| MFCD00000938 | |
| UN2922 | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| COC(=O)CC(=O)CCl | |
| 150.56 | |
| 150.56 | |
| Liquid |
Sécurité et manipulation
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 251-230-5
missing translation for 'rtecsNumber' : AK5115000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only