Learn More
Thermo Scientific Chemicals Meloxicam, 99-101%
CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol
$100.63 - $100.63
Identifiants chimiques
| CAS | 71125-38-7 |
|---|---|
| Molecular Formula | C14H13N3O4S2 |
| Molecular Weight (g/mol) | 351.40 |
| MDL Number | MFCD00868752 |
| InChI Key | DWMREKMVXIFPFM-ACCUITESSA-N |
| Synonym | meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin |
| PubChem CID | 54677470 |
| ChEBI | CHEBI:6741 |
| IUPAC Name | 4-hydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1$l^{6},2-benzothiazine-3-carboxamide |
| SMILES | CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC459550050
|
Thermo Scientific Chemicals
459550050 |
5 g |
chaque for $100.63
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 71125-38-7 | |
| 351.40 | |
| DWMREKMVXIFPFM-ACCUITESSA-N | |
| 54677470 | |
| 4-hydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1$l^{6},2-benzothiazine-3-carboxamide |
| C14H13N3O4S2 | |
| MFCD00868752 | |
| meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin | |
| CHEBI:6741 | |
| CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O |
Spécifications
| 71125-38-7 | |
| 0.1% max. | |
| Authentic | |
| MFCD00868752 | |
| meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin | |
| DWMREKMVXIFPFM-ACCUITESSA-N | |
| 4-hydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1$l^{6},2-benzothiazine-3-carboxamide | |
| 54677470 | |
| 351.41 | |
| Meloxicam |
| 255.0°C | |
| 0.5% max. (105°C, 4 hr) | |
| C14H13N3O4S2 | |
| 5 g | |
| Solubility in water: 7,5g/L. Other solubilities: 5g/L dmso | |
| CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O | |
| 351.40 | |
| CHEBI:6741 | |
| 99 to 101% |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
RUO – Research Use Only