missing translation for 'onlineSavingsMsg'
Learn More

Meldrum's Acid (=2,2-Dimethyl-1,3-dioxane-4,6-dione) 98.0+%, TCI America™

Supplier:  TCI America M0799100G

 Afficher plus de versions de ce produit

Numéro de catalogue. M0799100G


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Meldrum′s Acid (=2,2-Dimethyl-1,3-dioxane-4,6-dione)
95°C
MFCD00006638
meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
CC1(OC(=O)CC(=O)O1)C
144.126
144.13
Crystalline Powder
2033-24-1
C6H8O4
100 g
GXHFUVWIGNLZSC-UHFFFAOYSA-N
2,2-dimethyl-1,3-dioxane-4,6-dione
16249
≥98.0% (T)

Identifiants chimiques

2033-24-1
144.126
GXHFUVWIGNLZSC-UHFFFAOYSA-N
16249
CC1(OC(=O)CC(=O)O1)C
C6H8O4
MFCD00006638
meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
2,2-dimethyl-1,3-dioxane-4,6-dione
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (5)-6991

missing translation for 'rtecsNumber' : JH0800000

missing translation for 'tsca' : Yes

Recommended Storage : Refrigerator

FDS
Documents

Documents

Promotions

Promotions