missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCP-1 ProQuantum Immunoassay Kit

Numéro de catalogue. A355985
Encompass
Modifier l'affichage
Click to view available options
Quantité:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A355985 5 x 96 Tests
A35598 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A355985 Fournisseur Invitrogen™ Code fournisseur A355985

ELISA

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.

Spécifications

Plage de l'essai 0.064 to 5000
Sensibilité d’essai <0.05 pg/mL
Conjugué FAM™
Méthode de détection Taqman™ Real-time PCR
Format de la trousse d’immunodosage Proximity-based Immunoassay
Type de produit ProQuantum
Type d’échantillon Plasma, Serum, Supernatant
Espèces cibles Human
À utiliser avec (application) Protein Quantitation
À utiliser avec (équipement) qPCR Instruments
ID du gène (Entrez) 6347
Temps d’incubation 1 hr.
Contenus des kits Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Étiquette ou colorant FAM™
Quantité 5 x 96 Tests
Statut réglementaire RUO
Volume d’échantillon Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Conditions de stockage Frozen at -20°C
Cible CCL2 (MCP-1)
Durée du test 30 mins
Durée totale de l'essai 2 hrs
Afficher plus de résultats Afficher moins de résultats