missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ Technic™ NS-450 Heavyweight Neoprene/Natural Rubber Gloves, Flock Lining

Gloves provide multiple layer protection with natural rubber inner layer for flexibility and tear strength, and neoprene outer layer for mechanical and chemical protection.

Supplier:  MAPA™ 450448

Numéro de catalogue. 11-392-36C



Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Multiple layer construction of neoprene outer layer and inner natural rubber
  • Embossed nonslip diamond grip for handling wet or greasy objects
  • Hand contoured for greater comfort
  • Cotton flock lining absorbs perspiration, maximizes easy on and off
  • Black

Spécifications

Spécifications

21 CFR 177-190
No
Large
0.76 mm
Flock
450428
16 in.
Black
Neoprene, Latex
30 mil
Diamond
Lined
Technic™
40.5 cm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions