missing translation for 'onlineSavingsMsg'
Learn More

Magnesium pyrophosphate

CAS: 13446-24-7 | Mg2O7P2 | 222.551 g/mol

122,85 $ - 993,86 $

Identifiants chimiques

CAS 13446-24-7
Molecular Formula Mg2O7P2
Molecular Weight (g/mol) 222.551
MDL Number MFCD00049482
InChI Key XZTWHWHGBBCSMX-UHFFFAOYSA-J
Synonym magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
PubChem CID 122213
IUPAC Name dimagnesium;phosphonato phosphate
SMILES [O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA3931718
Afficher les documents
Thermo Scientific Chemicals
03931718
50 g N/A
Il en reste null
 
AA3931730
Afficher les documents
Thermo Scientific Chemicals
03931730
250 g N/A
Il en reste null
 
AA39317A1
Afficher les documents
Thermo Scientific Chemicals
039317A1
1 kg N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

13446-24-7
222.551
XZTWHWHGBBCSMX-UHFFFAOYSA-J
122213
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]
Mg2O7P2
MFCD00049482
magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
dimagnesium;phosphonato phosphate

Spécifications

White
Powder
Mg2O7P2
14,5684
Soluble in dilute mineral acids. Insoluble in water.
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]
222.551
222.57
13446-24-7
50 g
MFCD00049482
magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
XZTWHWHGBBCSMX-UHFFFAOYSA-J
dimagnesium;phosphonato phosphate
122213
Magnesium pyrophosphate
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-595-0

TSCA : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

RUO – Research Use Only