Learn More
Lithium hydroxide monohydrate, ACS, 98% min
CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Supplier: Thermo Scientific Chemicals 04317136
Description
Lithium chloride is useful for the production of lithium metal and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Spécifications
| Lithium hydroxide monohydrate | |
| White | |
| 500 g | |
| H3LiO2 | |
| lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| lithium;hydroxide;hydrate | |
| 168937 | |
| ACS | |
| White crystalline powder |
| 1310-66-3 | |
| Powder | |
| 98% min. | |
| MFCD00149772 | |
| Soluble in water,methanol and ethanol. | |
| [Li+].O.[OH-] | |
| 41.96 | |
| 41.96 (23.95 Anhydrous) | |
| Air sensitive; Hygroscopic | |
| 1.51 g/cm3 at 20°C |
Identifiants chimiques
| 1310-66-3 | |
| 41.96 | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| 168937 |
| H3LiO2 | |
| MFCD00149772 | |
| lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
| [Li+].O.[OH-] |
Sécurité et manipulation
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 215-183-4
missing translation for 'rtecsNumber' : OJ6307080
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only