missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium dodecylsulfate, 98%

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol

$78.19 - $714.95

Identifiants chimiques

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
PubChem CID 2735071
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAA1600506
Afficher les documents
Thermo Scientific Chemicals
A1600506
5 g
chaque for $78.19
Il en reste null
 
AAA1600514
Afficher les documents
Thermo Scientific Chemicals
A1600514
25 g
chaque for $261.26
Il en reste null
 
AAA1600522
Afficher les documents
Thermo Scientific Chemicals
A1600522
100 g
chaque for $714.95
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

2044-56-6
272.33
YFVGRULMIQXYNE-UHFFFAOYSA-M
2735071
C12H25LiO4S
MFCD00007467
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Spécifications

2044-56-6
C12H25LiO4S
UN1325
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium(1+) dodecyl sulfate
2735071
98%
Lithium dodecylsulfate
5 g
MFCD00007467
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
272.33
272.33
Odorless
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H318-H228-H335-H315
Causes serious eye damage.
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 218-058-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only