missing translation for 'onlineSavingsMsg'
Learn More

Lithium arsenate, 99%

CAS: 13478-14-3 | AsLi3O4 | 159.738 g/mol

$94.62 - $307.16

Identifiants chimiques

CAS 13478-14-3
Molecular Formula AsLi3O4
Molecular Weight (g/mol) 159.738
MDL Number MFCD00053998
InChI Key VKJKOXNPYVUXNC-UHFFFAOYSA-K
Synonym lithium arsenate, trilithium arsenate, trilithium arsorate, arsenic acid h3aso4 , trilithium salt, acmc-20akn0, aso4.3li, arsenic acid h3aso4 , lithium salt 1:3, trilithium 1+ ion arsenate, arsenic acid h3aso4 ,lithium salt 1:3
PubChem CID 166828
IUPAC Name trilithium;trioxido(oxo)-$l^{5}-arsane
SMILES [Li+].[Li+].[Li+].[O-][As](=O)([O-])[O-]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA3935006
Afficher les documents
Thermo Scientific Chemicals
03935006
5 g
chaque for $94.62
Il en reste null
 
AA3935014
Afficher les documents
Thermo Scientific Chemicals
03935014
25 g
chaque for $307.16
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

13478-14-3
159.738
VKJKOXNPYVUXNC-UHFFFAOYSA-K
166828
[Li+].[Li+].[Li+].[O-][As](=O)([O-])[O-]
AsLi3O4
MFCD00053998
lithium arsenate, trilithium arsenate, trilithium arsorate, arsenic acid h3aso4 , trilithium salt, acmc-20akn0, aso4.3li, arsenic acid h3aso4 , lithium salt 1:3, trilithium 1+ ion arsenate, arsenic acid h3aso4 ,lithium salt 1:3
trilithium;trioxido(oxo)-$l^{5}-arsane

Spécifications

13478-14-3
5 g
MFCD00053998
lithium arsenate, trilithium arsenate, trilithium arsorate, arsenic acid h3aso4 , trilithium salt, acmc-20akn0, aso4.3li, arsenic acid h3aso4 , lithium salt 1:3, trilithium 1+ ion arsenate, arsenic acid h3aso4 ,lithium salt 1:3
VKJKOXNPYVUXNC-UHFFFAOYSA-K
trilithium;trioxido(oxo)-$l^{5}-arsane
166828
99%
Lithium arsenate
Powder
AsLi3O4
UN1557
Soluble in dilute acetic acid; Insoluble in water
[Li+].[Li+].[Li+].[O-][As](=O)([O-])[O-]
159.738
159.74
3.07 g/mL
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 236-773-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only