Learn More
Greiner Bio-One Leucosep Polypropylene Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
Supplier:  Greiner Bio-One 227288
 Afficher plus de versions de ce produit
Numéro de catalogue. 07000499
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Découvrez plus d'offres spéciales
Description
Description
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes
  • No recontamination with erythrocytes
  • Service Plan
    No
Spécifications
Spécifications
50 mL
Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow
Aseptically Manufactured
Centrifuge Tube
Polypropylene
Porous barrier
Conical
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS