Learn More
Abcam Inc anti-LAMP1, Polyclonal, Abcam™ DFS Item
Supplier:  Abcam Inc AB24170100UG
LAMP1, Host Species: Rabbit, Target Species: Bovine, Canine, Hamster, Human, Murine, Rat, Xenopus, Zebrafish, Polyclonal, Primary, 0.9-1mg/mL, Unconjugated, 100ug, Affinity Purified, Synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human LAMP1, IgG
Numéro de catalogue. NC9718638
Il en reste null
Découvrez plus d'offres spéciales