Learn More
Thermo Scientific Chemicals L-(-)-Arabitol, 98%
CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol
$93.49 - $1,008.01
Identifiants chimiques
| CAS | 7643-75-6 |
|---|---|
| Molecular Formula | C5H12O5 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00064290 |
| InChI Key | HEBKCHPVOIAQTA-UHFFFAOYNA-N |
| Synonym | l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol |
| PubChem CID | 439255 |
| ChEBI | CHEBI:18403 |
| IUPAC Name | (2S,4S)-pentane-1,2,3,4,5-pentol |
| SMILES | OCC(O)C(O)C(O)CO |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AAA1310306
|
Thermo Scientific Chemicals
A1310306 |
5 g |
chaque for $93.49
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA1310314
|
Thermo Scientific Chemicals
A1310314 |
25 g |
chaque for $325.56
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA1310322
|
Thermo Scientific Chemicals
A1310322 |
100 g |
chaque for $1,008.01
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).
Solubility
Soluble in water (50 mg/ml).
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Identifiants chimiques
| C5H12O5 | |
| MFCD00064290 | |
| l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol | |
| CHEBI:18403 | |
| OCC(O)C(O)C(O)CO |
Spécifications
| 7643-75-6 | |
| C5H12O5 | |
| 1720521 | |
| 14,762 | |
| Soluble in water (50mg/ml). | |
| OCC(O)C(O)C(O)CO | |
| (2S,4S)-pentane-1,2,3,4,5-pentol | |
| 439255 | |
| 152.15 | |
| L-(-)-Arabitol |
| 98°C to 103°C | |
| MFCD00064290 | |
| Hygroscopic | |
| l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| −11° (c=5 in 8% Borax Solution) | |
| 152.15 | |
| CHEBI:18403 | |
| 98% |
Sécurité et manipulation
H500
EINECSNumber : 231-582-6
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only